Läs mer
Tocris Bioscience™ PD 166285 dihydrochloride
GREENER_CHOICE
Produktkod. 18053972
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18053972 1 mg
18044402 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18053972 Leverantör Tocris Bioscience™ Leverantörsnummer 3785/1
Potent Src inhibitor; also inhibits FGFR1, PDGFRβ and Wee1
Potent inhibitor of the tyrosine kinases c-Src, fibroblast growth factor receptor 1 (FGFR1), and platelet-derived growth factor receptor β (PDGFRβ) (IC50 values are 8.4, 39.3 and 98.3 nM respectively). Also inhibits the checkpoint kinases Wee1 and Myt1; abolishes Cdc2 phosphorylation in numerous tumor cell lines and abrogates the G2 checkpoint.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 212391-63-4
Molekylformel C26H29Cl4N5O2
Specifikationer
Kemiskt namn eller material PD 166285 dihydrochloride
Kvantitet 1 mg
Formel vikt 585.35
Renhet >99%
Molekylvikt (g/mol) 585.351