Läs mer
Tocris Bioscience™ PD 334581
GREENER_CHOICE
Produktkod. 30001578
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001578 10 mg
30001598 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001578 Leverantör Tocris Bioscience™ Leverantörsnummer 4824/10
MEK1 inhibitor
Inhibitor of MEK1. Analog of PD 184352 (Cat. No. 4237).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 548756-68-9
Molekylformel C20H19F3IN5O2
Specifikationer
Kemiskt namn eller material PD 334581
Kvantitet 10 mg
Formel vikt 545.3
Renhet >98%
Molekylvikt (g/mol) 545.305