Läs mer
PD176252, MedChemExpress
Produktkod. 16446547
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16446547 5 mg
1 options
Produktkod. 16446547 Leverantör Medchem Express Leverantörsnummer HY103286
MedChemExpress PD176252 is a potent antagonist of neuromedin-B preferring (BB1) and gastrin-releasing peptide-preferring (BB2) receptor with Kis of 0.17 nM and 1 nM for human BB1 and BB2 receptors, and 0.66 nM, 16 nM for Rat BB1 and BB2 receptors, respectively; PD176252 is also an agonist of N-Formyl peptide receptor1/2 (FPR1/FPR2), with EC50s of 0.31 and 0.66 μM in HL-60 cells.
PD176252 is a potent antagonist of neuromedin-B preferring (BB1) and gastrin-releasing peptide-preferring (BB2) receptor with Kis of 0.17 nM and 1 nM for human BB1 and BB2 receptors, and 0.66 nM, 16 nM for Rat BB1 and BB2 receptors, respectively
Specifikationer
Kemiskt namn eller material PD176252
Fysisk form Powder
CAS 204067-01-6
Färg Yellow
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C32H36N6O5
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 115 mg/mL (196.69 mM; Need ultrasonic)
LEDER O=C(NC1=CC=C([N+]([O-])=O)C=C1)N[C@](CC2=CNC3=C2C=CC=C3)(C)C(NCC4(C5=CC=C(OC)C=N5)CCCCC4)=O
Molekylvikt (g/mol) 584.67
Formel vikt 584.67
Procent renhet 98.17%