Läs mer
Tocris Bioscience™ PDD 00017273
Beskrivning
PDD 00017273 is a potent and selective poly (ADP ribose) glycohydrolase (PARG) inhibitor (IC50 = 26 nM). Exhibits >350-fold selectivity for PARG over a panel of ion channels, enzymes and receptors, including PARP1 and ARH3. Maintains PAR chains and induces DNA double-stranded breaks in cells following DNA damage. Decreases colony formation of ZR-75-1 BRCA1 WT cells and inhibits cancer cell survival. Cell permeable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-1,2,3,4-tetrahydro-N-(1-methylcyclopropyl)-3-[(2-methyl-5-thiazolyl)methyl]-2,4-dioxo-6-quinazolinesulfonamide |
| CAS | 1945950-21-9 |
| Kvantitet | 10 mg |
| Löslighet | Soluble to 100mM in water and to 100mM in DMSO |
| Mål | PARG Inhibitors |
| Molekylformel | C23H26N6O4S2 |
| Formel vikt | 514.62 |
| Fysisk form | Solid |
| Färg | White |
| Produkttyp | Bioactive Small Molecule |
| Visa mer |