missing translation for 'onlineSavingsMsg'
Learn More

PDE4DIP Recombinant Protein, Abnova

Brand:  Abnova H00009659-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16183982

  • 5030.41 SEK / 25µg

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH26270.1
Solution
9659
PDE4DIP (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PDE4DIP
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
45.21
8
Glutathione Sepharose 4 Fast Flow
25 ug
MFLCQGFRKYLPEHLNDLKKENFSLKLRIYFLEERMQQKYEASREDIYRRNTELKVEVESLKRELQDKKQHLDKTWADVENLNSQNEAELRRQFEERQQETEHVYELLENKMQLLQEESRLAKNEAARMAALVEAEKECNLELSEKLKGVTKNWEDVPGDQVKPDQYTEALAQRDKI
CMYA2/DKFZp781J054/MGC75440/MMGL
PDE4DIP
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates