missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24842-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Removes the formyl group from the N-terminal Met of newly synthesized proteins.Specifications
Polyclonal | |
Unconjugated | |
PDF, PDF1A, Polypeptide deformylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64146 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HBH1 | |
PDF Fusion Protein Ag18233 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |