missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16823345
|
Proteintech
24842-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16813345
|
Proteintech
24842-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
Removes the formyl group from the N-terminal Met of newly synthesized proteins.Especificaciones
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9HBH1 | |
64146 | |
PDF Fusion Protein Ag18233 | |
Primary | |
-20°C | |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PDF, PDF1A, Polypeptide deformylase | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |