missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDIA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PDIA6 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16878294
|
Proteintech
18233-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16868294
|
Proteintech
18233-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to PDIA6.
P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.Specifications
PDIA6 | |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q15084, Q63081, Q922R8 | |
10130, 286906, 71853 | |
PDIA6 Fusion Protein Ag12940 | |
Primary | |
-20°C | |
PDIA6 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ER protein 5, ERP5, P5, PDIA6, Protein disulfide isomerase A6, Protein disulfide isomerase P5, TXNDC7 | |
Pdia6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |