missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PDK-1, His Tag Recombinant Protein
Human PDK-1, His Tag Recombinant Protein
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_002604 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 66.9 kDa |
Gene Alias | PDK-1 |
KinaseFamily | PDHK Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15478393
|
Invitrogen™
PR4194C |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10104392
|
Invitrogen™
PV4033 |
1 mg |
79000.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PDK1 plays a vital role in insulin-induced glycogen synthesis, PKC activation, and activation of the AGC subfamily of protein kinases.
Specifications
NP_002604 | |
66.9 kDa | |
PDHK Kinase Family | |
His Tag | |
Dry Ice |
Kinase Assay | |
PDK-1 | |
Human | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% Triton X-100, 5mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus |