Learn More
PDZD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546583
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PDPWSLERPR FCLLSKEEGK SFGFHLQQEL GRAGHVVCRV DPGTSAQRQG Sequence homology: Cow: 79%; Dog: 93%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 86%; Rat: 93%
PDZD3 Polyclonal antibody specifically detects PDZD3 in Human samples. It is validated for Western Blot.Specifications
PDZD3 | |
Polyclonal | |
Unconjugated | |
Pdzd3 | |
DLNB27; hypothetical protein LOC539171; IKEPP; intestinal and kidney enriched PDZ protein; intestinal and kidney-enriched PDZ protein; Na(+)/H(+) exchange regulatory cofactor NHE-RF4; na/Pi cotransporter C-terminal-associated protein 2; NaPi-Cap2; natrium-phosphate cotransporter IIa C-terminal-associated protein 2; Nherf4; NHERF-4; PDZ domain containing 2; PDZ domain containing 3; PDZ domain-containing protein 2; PDZ domain-containing protein 3; Pdzd3; Pdzk2; RGD1559807; Sodium-hydrogen exchanger regulatory factor 4; sodium-phosphate cotransporter IIa C-terminal-associated protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79849 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86UT5 | |
Pdzd3 | |
Synthetic peptide directed towards the N-terminal of human PDZD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |