missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18795-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Acts as a regulatory protein that associates with GUCY2C and negatively modulates its heat-stable enterotoxin-mediated activation.Specifications
PDZD3 | |
Polyclonal | |
Unconjugated | |
PDZD3 | |
IKEPP, NaPi Cap2, NHERF 4, NHERF4, PDZ domain containing 3, PDZD3, PDZK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170761, 500986, 79849 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86UT5, Q99MJ6 | |
Pdzd3 | |
PDZD3 Fusion Protein Ag6931 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |