missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PDZK1 (Human) Recombinant Protein
Human PDZK1 full-length ORF ( AAH06496, 1 a.a. - 519 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH06496 |
---|---|
Gene ID (Entrez) | 5174 |
Name | PDZ domain containing 1 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178901
|
Abnova™
H00005174-P01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16188901
|
Abnova™
H00005174-P01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MTSTFNPRECKLSKQEGQNYGFFLRIEKDTEGHLVRVVEKCSPAEKAGLQDGDRVLRINGVFVDKEEHMQVVDLVRKSGNSVTLLVLDGDSYEKAVKTRVDLKELGQSQKEQGLSDNILSPVMNGGVQTWTQPRLCYLVKEGGSYGFSLKTVQGKKGVYMTDITPQGVAMRAGVLADDHLIEVNGENVEDASHEEVVEKVKKSGSRVMFLLVDKETDKRHVEQKIQFKRETASLKLLPHQPRIVEMKKGSNGYGFYLRAGSEQKGQIIKDIDSGSPAEEAGLKNNDLVVAVNGESVETLDHDSVVEMIRKGGDQTSLLVVDKETDNMYRLAHFSPFLYYQSQELPNGSVKEAPAPTPTSLEVSSPPDTTEEVDHKPKLCRLAKGENGYGFHLNAIRGLPGSFIKEVQKGGPADLAGLEDEDVIIEVNGVNVLDEPYEKVVDRIQSSGKNVTLLVCGKKAYDYFQAKKIPIVSSLADPLDTPPDSKEGIVVESNHDSHMAKERAHSTASHSSSNSEDTEM
Specifications
AAH06496 | |
PDZ domain containing 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
CAP70, CLAMP, NHERF3, PDZD1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5174 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PDZK1 | |
GST |