missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEA15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21446-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Blocks Ras-mediated inhibition of integrin activation and modulates the ERK MAP kinase cascade. Inhibits RPS6KA3 activities by retaining it in the cytoplasm. Inhibits both TNFRSF6- and TNFRSF1A-mediated CASP8 activity and apoptosis. Regulates glucose transport by controlling both the content of SLC2A1 glucose transporters on the plasma membrane and the insulin-dependent trafficking of SLC2A4 from the cell interior to the surface.Specifications
PEA15 | |
Polyclonal | |
Unconjugated | |
PEA15 | |
HMAT1, HUMMAT1H, MAT1, MAT1H, PEA 15, PEA15, PED | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18611, 8682 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15121, Q62048 | |
Pea15 | |
PEA15 Fusion Protein Ag13782 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |