Learn More
Gilson™ PEEK Color-Coded High-Pressure Tubing
Brand: Gilson™ 02006-08
Additional Details : Weight : 0.25000kg
Specifications
High Pressure Tubing | |
Black | |
±0.001 in. | |
Smooth | |
1.58 mm | |
PEEK | |
-51°C |
Autoclavable | |
0 to 14 | |
Ethylene Oxide, Gamma Irradiation, Thermal | |
Great for High Pressure Applications | |
1.5 m | |
500 to 10,000 psi | |
100°C |