Learn More
GE Healthcare PEEK Male Connector
Connect tubing of different sizes. GE Healthcare Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Brand:  GE Healthcare 18-1120-93
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11320252
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • Chromatography accessory
  • Complete Packsize 2
  • Select from a variety of connections sizes and internal diamters 
  • For use with chromatography tubing
  • Connectors made of Polyetheretherketone (PEEK)
Specifications
Specifications
Male Connector
0.5 mm
1/16 in. Male to 1/16 in. Male
1/16 in. O.D. Tubing
PEEK