Learn More
Cytiva (Formerly GE Healthcare Life Sciences) PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Brand:  Cytiva (Formerly GE Healthcare Life Sciences) 18-1120-93
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11320252
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Chromatography accessory
  • Complete Packsize 2
  • Select from a variety of connections sizes and internal diamters 
  • For use with chromatography tubing
  • Connectors made of Polyetheretherketone (PEEK)
Specifications
Specifications
0.5 mm
PEEK
1/16 in. Male to 1/16 in. Male
1/16 in. O.D. Tubing
Male Connector