Learn More
Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 100 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
5115.00 SEK
Specifications
Diameter (English) Outer | 1/32 in. |
---|---|
Overall Length (English) | 100 ft. |
Material | PEEK |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
16622552
|
Trajan Scientific and Medical
1301002500-100F |
Natural | 0.0025 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
16662552
|
Trajan Scientific and Medical
1301004006-100F |
Black | 0.004 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15770979
|
Trajan Scientific and Medical
1301005001-100F |
Red | 0.005 in. |
5115.00 SEK
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
15710989
|
Trajan Scientific and Medical
1301007002-100F |
Yellow | 0.007 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15750989
|
Trajan Scientific and Medical
1301010003-100F |
Blue | 0.010 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15790989
|
Trajan Scientific and Medical
1301015008-100F |
Gray | 0.015 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
16602562
|
Trajan Scientific and Medical
1301015000-100F |
Natural | 0.015 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15730999
|
Trajan Scientific and Medical
1301020004-100F |
Orange | 0.020 in. |
5115.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
1/32 in. | |
100 ft. | |
PEEK | |
100°C |
For research use only