Läs mer
PEG-2 Hydrogenated Tallow Amine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tallowamine polyethoxylated |
| CAS | 61791-26-2 |
| Kvantitet | 5 g |
| Synonym | Amines','tallow alkyl','ethoxylated, Agnique Tam 5, Alkaminox T 12, Alkaminox T 2, Amiet 502, Amiladin D, Amines','tallow','ethoxylated, Amines','tallow','polyethoxylated, Atlas G 3762, Atlas G 3780644, Atlas G 3780A, Atmer 261, Berol 387, Berol 392, Chemeen T 5, Crisomin T 15, Crodamet T 15, Emulson AG/GPE 3CA, Emulson AG/GPE 3SSM, Entry II, Ethokem, Ethomeen T, Ethomeen T 105, Ethomeen T 12, Ethomeen T 12E, Ethomeen T 15, Ethomeen T 16, Ethomeen T 18, Ethomeen T 20, Ethomeen T 25, Ethomeen T 27, Ethomeen T 30, Ethomeen T 40, Ethomeen T 70, Ethomeen YT, Ethox TAM 15, Ethox TAM 2, Ethox TAM 20, Ethox TAM 5, Ethoxamine SF 11, Ethoxamine SF 15, Ethoxylated tallow alkyl amines, Ethoxylated tallow amines, Ethylan TT 05, Ethylan TT 15, Ethylan TT 203, Ethylan TT 40, Fentacare T 15, Frigate, G 3780A, Genamin 267, Genamin T, Genamin T 020, Genamin T 050, Genamin T 080, Genamin T 100, Genamin T 120, Genamin T 150, Genamin T 200, Genamin T 200NF, Genamin T 250, Hostastat FA 38, Hyspray, Icomeen T 15, Icomeen T 2, Imbentin TAM 400, Jeetox T 2, KFG 597C, Katapol PN 430, Katapol PN 530, Katapol PN 730, Katapol VP 532, Kemamine AS 974-1, MON 0818, Mazeen T 2, Meteor, Nalco NM 159, Newcol TA 420, Newkalgen D 3615T, Nissan Nymeen T 2-202, Nissan Nymeen T 2-210, Nissan Nymeen T 2-230, Nissan Nymeen T 2-260, Noramox S, Noramox S 1, Noramox S 11, Noramox S 2, Noramox S 5, Noramox S 7, Nymeen T 2-202, Nymeen T 2-210, Nymeen T 2-230, Prevocell 1618/3, Rhodameen IT 50/46, Rhodameen PN 430, Rhodameen RAM 7, Rhodameen T 12/90, Rhodameen T 15, Rhodameen T 50, Rhodameen T 7, Rhodameen VP 532, Rhodameen VP 532SPB, Surfonic AGM 510, Surfonic T 10, Surfonic T 15, Surfonic T 2, Surfonic T 20, Surfonic T 5, Surfonic T 6, T 15, TAM 105, Tallow alkyl amines','ethoxylated, Tam 15, Teric 17M2, Teric 204, Terwet 3780, Terwet 3784, Toximul 8362, Toximul TA 10, Toximul TA 15, Toximul TA 2, Toximul TA 5, Toximul TA 6, Trymeen 6606, Trymeen 6607, Trymeen TAM, VP 532SPB, Varonic T 202, Varonic T 205, Varonic T 210, Varonic T 216, Varonic T 220, Varonic T 220D, Weedmaster CT Surfactant, Witcamine 405, Witcamine 6606, Witcamine TAM 105, Witcamine TAM 45, Witconol TD 1407 |