Läs mer
Peiminine, TRC
Produktkod. 30416725
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30416725 10 mg
1 options
Produktkod. 30416725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP21838010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Peiminine
CAS 18059-10-4
Molekylformel C27 H43 N O3
Kvantitet 10 mg
Synonym Cevan-6-one','3,20-dihydroxy-','(3β,5α)-, 5α-Cevan-6-one','3β,20-dihydroxy- (8CI), Fritillarine (6CI), Osnovanine (7CI), Benzo[7,8]fluoreno[2,1-b]quinolizine','cevan-6-one deriv., (3β,5α)-3,20-Dihydroxycevan-6-one, Peiminine, Raddeanine, Raddeanine (Fritillaria), Verticinone
InChI formel InChI=1S/C27H43NO3/c1-15-4-7-25-27(3,31)21-6-5-17-18(20(21)14-28(25)13-15)11-22-19(17)12-24(30)23-10-16(29)8-9-26(22,23)2/h15-23,25,29,31H,4-14H2,1-3H3/t15-,16-,17+,18+,19-,20-,21-,22-,23+,25-,26+,27-/m0/s1
LEDER C[C@@]12[C@@]3([C@]([C@]4([C@@](C3)([C@]5([C@](CC4)([C@](C)(O)[C@]6(N(C5)C[C@@H](C)CC6)[H])[H])[H])[H])[H])(CC(=O)[C@]1(C[C@@H](O)CC2)[H])[H])[H]
IUPAC-namn (3S,4aS,6aS,6bS,8aS,9S,9aS,12S,15aS,15bR,16aS,16bR)-3,9-dihydroxy-9,12,16b-trimethyldocosahydrobenzo[4,5]indeno[1,2-h]pyrido[1,2-b]isoquinolin-5(1H)-one
Molekylvikt (g/mol) 429.65
Formel vikt 429.3243