missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PELI2 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895584
|
Proteintech
16097-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16885584
|
Proteintech
16097-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 19-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Scaffold protein which probably links Toll-like receptors (TLRs) to basic cellular processes via its interaction with the complex containing IRAK kinases and TRAF6. Can activate the MAP (mitogen activated protein) kinase pathway leading to activation of ELK1. Not required for NF-kappa-B activation.Specifications
PELI2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8BST6, Q9HAT8 | |
305835, 57161, 93834 | |
PELI2 Fusion Protein Ag8940 | |
Primary | |
-20°C | |
PELI2 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
E3 ubiquitin-protein ligase pellino homolog 2, PELI2, Pellino 2, pellino homolog 2 (Drosophila), Protein pellino homolog 2 | |
PELI2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |