Learn More
3M™ PELTOR™ Optime™ Push To Listen Earmuffs
3M™ PELTOR™ Optime™ Push To Listen Earmuffs are electronic passive earmuffs, which have one passive side and one electronic side.
3910.00 SEK - 4350.00 SEK
Specifications
Product Type Earmuffs
View More Specs
Products 3
Product Code Brand Color Noise Reduction Rating (NRR) Price Quantity & Availability  
Product Code Brand Color Noise Reduction Rating (NRR) Price Quantity & Availability  
15739727 View Documents 3M™
7000039734
Black 29 dB 4175.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
15729727 View Documents 3M™
7000039733
Black 31 dB 3910.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
15799987 View Documents 3M™
7000108448
Hi-Viz 31 dB 4350.00 SEK
Pack of 10 View alternative products
Save Up to 
»
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Hearing protection with a manual listening function, simply push the button to listen, opening up the ear protector for communication
  • Pressing a button on the shell enables you to reduce attenuation level to follow a conversation
  • The electronic function automatically shuts off after 30 seconds and can be manually adjusted to close earlier if needed
Specifications
Specifications
Earmuffs