Läs mer
Pemetrexed acid, TRC
Produktkod. 30317250
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30317250 100 mg
1 options
Produktkod. 30317250 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA604905100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid
CAS 137281-39-1
Molekylformel C15 H14 N4 O3
Kvantitet 100 mg
Synonym Benzoic acid, 4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]-, 4-[2-(2-Amino-4-oxo-3,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid, 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid, Pemetrexed acid, Benzoic acid, 4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]-, 4-[2-(2-Amino-4-oxo-3,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid
InChI formel InChI=1S/C15H14N4O3/c16-15-18-12-11(13(20)19-15)10(7-17-12)6-3-8-1-4-9(5-2-8)14(21)22/h1-2,4-5,7H,3,6H2,(H,21,22)(H4,16,17,18,19,20)
LEDER NC1=Nc2[nH]cc(CCc3ccc(cc3)C(=O)O)c2C(=O)N1
IUPAC-namn 4-[2-(2-amino-4-oxo-3,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid
Molekylvikt (g/mol) 298.3
Formel vikt 298.1066