Läs mer
Pemetrexed, MedChemExpress
Produktkod. 16475136
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16475136 100 mg
16460527 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16475136 Leverantör Medchem Express Leverantörsnummer HY10820
MedChemExpress Pemetrexed (LY231514) is an antifolate, the Ki values of the pentaglutamate of Pemetrexed (LY231514) are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.
Pemetrexed (LY231514) is an antifolate, the Ki values of the pentaglutamate of Pemetrexed (LY231514) are 1.3, 7.2, and 65 nM for inhibits thymidylate synthase (TS), dihydrofolate reductase (DHFR), and glycinamide ribonucleotide formyltransferase (GARFT), respectively.
Specifikationer
Kemiskt namn eller material Pemetrexed
CAS 137281-23-3
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C20H21N5O6
Kvantitet 100 mg
Synonym LY231514
Löslighetsinformation DMSO : 250 mg/mL (584.92 mM; Need ultrasonic)
LEDER O=C1NC(N)=NC2=C1C(CCC3=CC=C(C(N[C@H](C(O)=O)CCC(O)=O)=O)C=C3)=CN2
Molekylvikt (g/mol) 427.41
Formel vikt 427.41
Procent renhet 98.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death