Läs mer
Pemetrexed Methyl Ester, TRC
Produktkod. 30412916
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30412916 10 mg
1 options
Produktkod. 30412916 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP21952510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pemetrexed Dimethyl Ester
CAS 155405-81-5
Molekylformel C22 H25 N5 O6
Kvantitet 10 mg
Synonym L-Glutamic acid','N-[4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-','1,5-dimethyl ester, L-Glutamic acid','N-[4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-','dimethyl ester (9CI), 1H-Pyrrolo[2,3-d]pyrimidine','L-glutamic acid deriv.
InChI formel InChI=1S/C22H25N5O6/c1-32-16(28)10-9-15(21(31)33-2)25-19(29)13-6-3-12(4-7-13)5-8-14-11-24-18-17(14)20(30)27-22(23)26-18/h3-4,6-7,11,15H,5,8-10H2,1-2H3,(H,25,29)(H4,23,24,26,27,30)/t15-/m0/s1
LEDER COC(=O)CC[C@H](NC(=O)c1ccc(CCc2c[nH]c3NC(=NC(=O)c23)N)cc1)C(=O)OC
IUPAC-namn dimethyl (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate
Molekylvikt (g/mol) 455.4638
Formel vikt 455.1805