Läs mer
Penciclovir Diacetate, TRC
Produktkod. 30411619
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30411619 50 mg
1 options
Produktkod. 30411619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP22150550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Penciclovir Diacetate
CAS 97845-72-2
Molekylformel C14 H19 N5 O5
Kvantitet 50 mg
Synonym 9-[4-(acetyloxy)-3-[(acetyloxy)methyl]butyl]-2-amino-1,9-dihydro-6H-Purin-6-one, BRL 39913, BRU 39913, Di-O-acetylpenciclovir, Penciclovir diacetate, O,O-Diacetylpenciclovir
InChI formel InChI=1S/C14H19N5O5/c1-8(20)23-5-10(6-24-9(2)21)3-4-19-7-16-11-12(19)17-14(15)18-13(11)22/h7,10H,3-6H2,1-2H3,(H3,15,17,18,22)
LEDER CC(=O)OCC(CCn1cnc2C(=O)N=C(N)Nc12)COC(=O)C
IUPAC-namn [2-(acetyloxymethyl)-4-(2-amino-6-oxo-3H-purin-9-yl)butyl] acetate
Molekylvikt (g/mol) 337.33
Formel vikt 337.1386