Learn More
Penciclovir Monoacetate, TRC
Product Code. 30428658
Change view
Click to view available options
Kvantitet:
10 mg
Product Code. Kvantitet
30428658 10 mg
1 options
Product Code. 30428658 Supplier Toronto Research Chemicals Supplier No. TRCP22151510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Penciclovir Monoacetate
CAS 97845-80-2
Molekylformel C12H17N5O4
Kvantitet 10 mg
Synonym BRL 42222, Mono-O-acetylpenciclovir, Penciclovir Monoacetate
InChI formel InChI=1S/C12H17N5O4/c1-7(19)21-5-8(4-18)2-3-17-6-14-9-10(17)15-12(13)16-11(9)20/h6,8,18H,2-5H2,1H3,(H3,13,15,16,20)
LEDER O=C1NC(N)=NC2=C1N=CN2CCC(CO)COC(C)=O
Molekylvikt (g/mol) 295.29
Formel vikt 295.1281