Läs mer
Pendimethalin, TRC
Produktkod. 30426016
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30426016 5 g
1 options
Produktkod. 30426016 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2219005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pendimethalin
CAS 40487-42-1
Molekylformel C13 H19 N3 O4
Kvantitet 5 g
Synonym N-(1-Ethylpropyl)-3,4-dimethyl-2,6-dinitrobenzenamine, 3,4-Dimethyl-2,6-dinitro-N-(1-ethylpropyl)aniline, 3,4-Dimethyl-2,6-dinitro-N-(1'-ethylpropyl)aniline, AC 92553, Accotab, Activus EC, Acumen, Gogosan, Herbadox, Most Micro, N-(1-Ethylpropyl)-2,6-dinitro-3,4-xylidine, N-(1-Ethylpropyl)-3,4-dimethyl-2,6-dinitroaniline, OWC, Ornamental Weedgrass Control, Panida, Panida 330EC, Pendimax, Pendimethalin, Pendimethaline, Pendulum, Penoxalin, Penoxaline, Phenoxalin, Prowl, Prowl 3.3, Prowl 400, SWGC, Southern Weed Grass Control, Stomp, Stomp 330, Stomp 330D, Stomp 330E, Stomp 330EC, Benzenamine','N-(1-ethylpropyl)-3,4-dimethyl-2,6-dinitro-
InChI formel InChI=1S/C13H19N3O4/c1-5-10(6-2)14-12-11(15(17)18)7-8(3)9(4)13(12)16(19)20/h7,10,14H,5-6H2,1-4H3
LEDER CCC(CC)Nc1c(cc(C)c(C)c1[N+](=O)[O-])[N+](=O)[O-]
IUPAC-namn 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline
Molekylvikt (g/mol) 281.31
Formel vikt 281.1376