Läs mer
Pendimethalin, TRC
Produktkod. 30417445
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30417445 500 mg
1 options
Produktkod. 30417445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP221900500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pendimethalin
CAS 40487-42-1
Molekylformel C13 H19 N3 O4
Kvantitet 500 mg
Synonym N-(1-Ethylpropyl)-3,4-dimethyl-2,6-dinitrobenzenamine, 3,4-Dimethyl-2,6-dinitro-N-(1-ethylpropyl)aniline, 3,4-Dimethyl-2,6-dinitro-N-(1'-ethylpropyl)aniline, AC 92553, Accotab, Activus EC, Acumen, Gogosan, Herbadox, Most Micro, N-(1-Ethylpropyl)-2,6-dinitro-3,4-xylidine, N-(1-Ethylpropyl)-3,4-dimethyl-2,6-dinitroaniline, OWC, Ornamental Weedgrass Control, Panida, Panida 330EC, Pendimax, Pendimethalin, Pendimethaline, Pendulum, Penoxalin, Penoxaline, Phenoxalin, Prowl, Prowl 3.3, Prowl 400, SWGC, Southern Weed Grass Control, Stomp, Stomp 330, Stomp 330D, Stomp 330E, Stomp 330EC, Benzenamine','N-(1-ethylpropyl)-3,4-dimethyl-2,6-dinitro-
InChI formel InChI=1S/C13H19N3O4/c1-5-10(6-2)14-12-11(15(17)18)7-8(3)9(4)13(12)16(19)20/h7,10,14H,5-6H2,1-4H3
LEDER CCC(CC)Nc1c(cc(C)c(C)c1[N+](=O)[O-])[N+](=O)[O-]
IUPAC-namn 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline
Molekylvikt (g/mol) 281.31
Formel vikt 281.1376