Läs mer
Penicillin G Benzathine Tetrahydrate, TRC
Produktkod. 30418621
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30418621 250 mg
1 options
Produktkod. 30418621 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP223410250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Penicillin G Benzathine Tetrahydrate
CAS 41372-02-5
Molekylformel C16 H20 N2 . 2 C16 H18 N2 O4 S . 4 H2 O
Kvantitet 250 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-[2S-(2α,5α,6β)]-','compd. with N,N'-bis(phenylmethyl)-1,2-ethanediamine (2:1)','tetrahydrate, 1,2-Ethanediamine','N,N'-bis(phenylmethyl)-','bis[(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate]','tetrahydrate (9CI), 1,2-Ethanediamine','N,N'-bis(phenylmethyl)-','bis[[2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate]','tetrahydrate
InChI formel InChI=1S/2C16H18N2O4S.C16H20N2.4H2O/c2*1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16;;;;/h2*3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);1-10,17-18H,11-14H2;4*1H2/t2*11-,12+,14-;;;;;/m11...../s1
LEDER O.O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(=O)O.CC4(C)S[C@@H]5[C@H](NC(=O)Cc6ccccc6)C(=O)N5[C@H]4C(=O)O.C(CNCc7ccccc7)NCc8ccccc8
IUPAC-namn N,N'-dibenzylethane-1,2-diamine;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;tetrahydrate
Molekylvikt (g/mol) 981.18
Formel vikt 980.4024