missing translation for 'onlineSavingsMsg'
Läs mer

Penicillin G kaliumsalt, 99,9 %, MP Biomedicals™

Product Code. p-4403784 Shop All MP Biomedicals Products
Click to view available options
Kvantitet:
1 mU
10 mU
100 mU
Unit Size:
1 mikroenhet
10 mikroenheter
100 mikroenheter
This item is not returnable. View return policy

Product Code. 11467600

Brand: MP Biomedicals 0210054301

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Chemical Identifiers

CAS 113-98-4
Molekylformel C33H36ClKN4O4S2
Molekylvikt (g/mol) 691.34
MDL-nummer MFCD00036193
InChI-nyckel HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin potassium salt
PubChem CID 23664709
ChEBI CHEBI:7963
IUPAC-namn kalium-[3-(2-klor-10H-fenotiazin-10-yl)propyl]dimetylamin (2S,5R,6R)-3,3-dimetyl-7-oxo-6-(2-fenylacetamido)-4-tia-1-azabicyklo[3.2.0]heptan-2-karboxylat
LEDER [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Kemiskt namn eller material Penicillin G potassium salt
Typ Penicillin G Potassium Salt
Namnnotering Potency: 1500-1700 u/mg
Fysisk form Pulver
Färg Vitt
Kvantitet 1 mU
Analysprocentintervall 0.999
Formel vikt 372.5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.