Läs mer
Penta-O-pivaloyl-Beta-D-glucopyranose, TRC
Produktkod. 30422603
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30422603 1 g
1 options
Produktkod. 30422603 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2301101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Penta-O-pivaloyl-beta-D-glucopyranose
CAS 81058-26-6
Molekylformel C31H52O11
Kvantitet 1 g
Synonym 1,2,3,4,6-Pentakis(2,2-dimethylpropanoate) β-D-Glucopyranose
InChI formel InChI=1S/C31H52O11/c1-27(2,3)22(32)37-16-17-18(39-23(33)28(4,5)6)19(40-24(34)29(7,8)9)20(41-25(35)30(10,11)12)21(38-17)42-26(36)31(13,14)15/h17-21H,16H2,1-15H3/t17-,18-,19+,20-,21+/m1/s1
LEDER CC(C)(C)C(O[C@@H]1[C@H]([C@@H](O[C@@H]([C@H]1OC(C(C)(C)C)=O)COC(C(C)(C)C)=O)OC(C(C)(C)C)=O)OC(C(C)(C)C)=O)=O
IUPAC-namn (2S,3R,4S,5R,6R)-6-(((2,2-dimethylpropanoyl)oxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrakis(2,2-dimethylpropanoate)
Molekylvikt (g/mol) 600.74
Formel vikt 600.35