Läs mer
Pentabromobenzyl Acrylate, TRC
Produktkod. 30421789
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30421789 100 mg
1 options
Produktkod. 30421789 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP227120100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pentabromobenzyl acrylate
CAS 59447-55-1
Molekylformel C10 H5 Br5 O2
Kvantitet 100 mg
Synonym 2-Propenoic acid','(2,3,4,5,6-pentabromophenyl)methyl ester, 2-Propenoic acid','(pentabromophenyl)methyl ester (9CI), 2,3,4,5,6-Pentabromobenzyl acrylate, Actimer FR 1025M, FR 1025M, PBBA
InChI formel InChI=1S/C10H5Br5O2/c1-2-5(16)17-3-4-6(11)8(13)10(15)9(14)7(4)12/h2H,1,3H2
LEDER Brc1c(Br)c(Br)c(COC(=O)C=C)c(Br)c1Br
IUPAC-namn (2,3,4,5,6-pentabromophenyl)methyl prop-2-enoate
Molekylvikt (g/mol) 556.6655
Formel vikt 551.6206