Learn More
Pentacarbonyliron, 99.5%, Thermo Scientific Chemicals
Catalog Number p-7049907
Click to view available options
Quantity:
25 g
13463-40-6
C5FeO5
195.90
MFCD00011001
FYOFOKCECDGJBF-UHFFFAOYSA-N
Iron pentacarbonyl; Iron carbonyl
pentakis(methanidylidyneoxidanium) iron
[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
13463-40-6
C5FeO5
195.90
MFCD00011001
FYOFOKCECDGJBF-UHFFFAOYSA-N
Iron pentacarbonyl; Iron carbonyl
pentakis(methanidylidyneoxidanium) iron
[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
As a catalyst in organic reactions, and as carbonyl iron for high-frequency coils
Pentacarbonyliron is used as a catalyst in organic reactions, and as carbonyl iron for high-frequency coils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -20°C
Color Red
Density 1.457 g/mL
Boiling Point 103°C (200°C (decomposition))
Flash Point -15°C (5°F)
Odor Pungent
Linear Formula Fe(CO)5
Refractive Index 1.5196
Quantity 25 g
UN Number UN1994
Sensitivity Air and light sensitive
Merck Index 14,5099
Solubility Information Soluble in nickel tetracarbonyl and organic solvents; Insoluble in water
Formula Weight 195.9
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Pentacarbonyliron
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentacarbonyliron
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Acute toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.