Läs mer
Pentachloronitrobenzene-13C6, TRC
Produktkod. 17960945
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960945 1 mg
17930505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17960945 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2379921MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 362044-64-2
Molekylformel 13C6 Cl5 N O2
Molekylvikt (g/mol) 301.29
Synonym Benzene-13C6, pentachloronitro- (9CI), Pentachloronitrobenzene (13C6), Pentachloronitrobenzene 13C6, Pentachloronitrobenzene-1,2,3,4,5,6-13C6, Quintozene-13C6
IUPAC-namn 1,2,3,4,5-pentachloro-6-nitro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene
LEDER [O-][N+](=O)[13c]1[13c](Cl)[13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl
Specifikationer
Kemiskt namn eller material Pentachloronitrobenzene-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C6Cl5NO2/c7-1-2(8)4(10)6(12(13)14)5(11)3(1)9/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 298.8573
Procent renhet >95%