Läs mer
Pentachlorophenol Sodium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pentachlorophenol sodium |
| CAS | 131-52-2 |
| Molekylformel | C6 Cl5 O . Na |
| Kvantitet | 2.5 g |
| Synonym | Phenol','pentachloro-','sodium deriv. (6CI), Phenol','pentachloro-','sodium salt (8CI,9CI), Biocel WD, CL 208, Dowicide G, GR 48-11PS, GR 48-32S, Mystox D, NAPCP, PCP sodium salt, PCP-Na, PCP-Sodium, PKhFN, Pentachlorophenate sodium, Pentachlorophenol sodium salt, Pentachlorophenoxy sodium, Pentanot 25, Preventol PN, Santobrite, Sapco 25, Sodium PCP, Sodium pentachlorophenate, Sodium pentachlorophenol, Sodium pentachlorophenolate, Sodium pentachlorophenoxide, Witophen N |
| InChI formel | InChI=1S/C6HCl5O.Na/c7-1-2(8)4(10)6(12)5(11)3(1)9;/h12H;/q;+1/p-1 |
| LEDER | [Na+].[O-]c1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl |
| IUPAC-namn | sodium;2,3,4,5,6-pentachlorophenolate |
| Molekylvikt (g/mol) | 288.32 |
| Formel vikt | 285.8289 |