Läs mer
Pentacosanoic Acid, TRC
Produktkod. 30416394
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30416394 100 mg
1 options
Produktkod. 30416394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP238193100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pentacosanoic Acid
CAS 506-38-7
Molekylformel C25 H50 O2
Kvantitet 100 mg
Synonym Pentacosanoic acid (6CI','7CI','8CI','9CI','ACI), Hyenic acid, n-Pentacosanoic acid, NSC 89289, Pentacosylic acid
InChI formel InChI=1S/C25H50O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25(26)27/h2-24H2,1H3,(H,26,27)
LEDER CCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
IUPAC-namn pentacosanoic acid
Molekylvikt (g/mol) 382.66
Formel vikt 382.3811