Läs mer
Pentadecanoic Acid 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl Ester, TRC
Produktkod. 30134421
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134421 250 mg
17912895 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134421 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP227140250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1404053-61-7
Molekylformel C38H55FN4O4
Molekylvikt (g/mol) 650.87
Synonym Paliperidone Pentadecanoate, 9-Hydroxyrisperidone Pentadecanoate
LEDER [Na+].O=C([O-])C1[C@H](O)C(O)C(O)[C@@H](O1)OC2=C(C)N(C)C=CC2=O
Specifikationer
Kemiskt namn eller material Pentadecanoic Acid 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl Ester
Kvantitet 250 mg
InChI formel InChI=1S/C13H17NO8.Na/c1-5-10(6(15)3-4-14(5)2)21-13-9(18)7(16)8(17)11(22-13)12(19)20;/h3-4,7-9,11,13,16-18H,1-2H3,(H,19,20);/q;+1/p-1/t7?,8-,9?,11?,13-;/m1./s1
Formel vikt 650.4207