Läs mer
Pentaerythritol Triacrylate (Technical Grade), TRC
Produktkod. 30137161
Change view
Kvantitet:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137161 25 g
17923815 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137161 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP23051525G
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3524-68-3
Molekylformel C14H18O7
Molekylvikt (g/mol) 298.29
Synonym SR 444, SR 444C, SR 444D, SR 444NS, SR 534, Sartomer 444, Sartomer SR 444, Setalux UV 2242, Light Acrylate DPE 3A, Light Acrylate PE 3A, Light Ester PE 3A, M 305, M 306, M 340, M 430, Miramer M 340, NK Ester A-TMM 3L New, NK Ester, A-TMN 3LM-N, NK Ester TMM 50T, Newfrontier PET 3, P 300, P 300 (Acrylate), PE 003, PE 3A, PET 3, PET 30, PET 3A, PETA, PETIA, PETRA;
IUPAC-namn [2-(hydroxymethyl)-3-prop-2-enoyloxy-2-(prop-2-enoyloxymethyl)propyl] prop-2-enoate
LEDER OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
Specifikationer
Kemiskt namn eller material Pentaerythritol Triacrylate (Technical Grade)
Kvantitet 25 g
InChI formel InChI=1S/C14H18O7/c1-4-11(16)19-8-14(7-15,9-20-12(17)5-2)10-21-13(18)6-3/h4-6,15H,1-3,7-10H2
Formel vikt 298.1053