Läs mer
Pentaerythritol Tris Ester With 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid, TRC
Produktkod. 30007772
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007772 10 mg
30011290 50 mg
30012323 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007772 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19429510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84633-54-5
Molekylformel C56 H84 O10
Molekylvikt (g/mol) 917.26
Synonym 1,1'-[2-[[3-[3,5-Bis(1,1-dimethylethyl)-4-hydroxyphenyl]-1-oxopropoxy]methyl]-2-(hydroxymethyl)-1,3-propanediyl] bis[3,5-bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoate], Pentaerythritol Tris Ester with 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid, 2-(((3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propanoyl)oxy)methyl)-2-(hydroxymethyl)propane-1,3-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)
IUPAC-namn [2,2-bis[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxymethyl]-3-hydroxypropyl] 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
LEDER CC(C)(C)c1cc(CCC(=O)OCC(CO)(COC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)COC(=O)CCc3cc(c(O)c(c3)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C
Specifications
Kemiskt namn eller material Pentaerythritol Tris Ester With 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C56H84O10/c1-50(2,3)38-25-35(26-39(47(38)61)51(4,5)6)19-22-44(58)64-32-56(31-57,33-65-45(59)23-20-36-27-40(52(7,8)9)48(62)41(28-36)53(10,11)12)34-66-46(60)24-21-37-29-42(54(13,14)15)49(63)43(30-37)55(16,17)18/h25-30,57,61-63H,19-24,31-34H2,1-18H3
Formel vikt 916.6064