Learn More
Pentafluorobenzonitrile, 99%, Thermo Scientific Chemicals
Catalog Number p-7077189
Click to view available options
Quantity:
5 g
25 g
100 g
773-82-0
C7F5N
193.07
YXWJGZQOGXGSSC-UHFFFAOYSA-N
pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k
69882
2,3,4,5,6-pentafluorobenzonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)F)F
773-82-0
C7F5N
193.07
YXWJGZQOGXGSSC-UHFFFAOYSA-N
pentafluorobenzonitrile, perfluorobenzonitrile, benzonitrile, pentafluoro, pentafluorocyanobenzene, benzonitrile, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro benzonitrile, 2,3,4,5,6-pentafluorobenzenecarbonitrile, cyanopentafluorobenzene, pubchem2319, acmc-209p9k
69882
2,3,4,5,6-pentafluorobenzonitrile
C(#N)C1=C(C(=C(C(=C1F)F)F)F)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 1.0°C to 3.0°C
Density 1.5300g/mL
Boiling Point 161.0°C to 162.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula C6F5CN
Refractive Index 1.4415 to 1.4435
Quantity 5 g
Specific Gravity 1.53
Solubility Information Solubility in water: insoluble.
Formula Weight 193.07
Percent Purity 99%
Chemical Name or Material Pentafluorobenzonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pentafluorobenzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.