missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl trifluoroacetate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7036266
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14533-84-7
C8F8O2
280.07
MFCD00134438
VCQURUZYYSOUHP-UHFFFAOYSA-N
pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8
4327891
(2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F
This item is not returnable. View return policy
14533-84-7
C8F8O2
280.07
MFCD00134438
VCQURUZYYSOUHP-UHFFFAOYSA-N
pentafluorophenyl trifluoroacetate, perfluorophenyl 2,2,2-trifluoroacetate, trifluoroacetic acid pentafluorophenyl ester, pentafluorphenyl trifluoracetate, pentafluorophenyltrifluoroacetate, pentafluorophenyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, pentafluorophenyl ester, acetic acid,2,2,2-trifluoro-, 2,3,4,5,6-pentafluorophenyl ester, ambotzrl-1046, acmc-1bui8
4327891
(2,3,4,5,6-pentafluorophenyl) 2,2,2-trifluoroacetate
FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 122.5°Cat760mmHg.

Notes
Hygroscopic. Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.63
Boiling Point 122°C to 123°C
Flash Point 52°C (126°F)
Refractive Index 1.368
Quantity 5 g
UN Number UN3272
Beilstein 2003848
Sensitivity Moisture sensitive
Solubility Information Soluble in water at 122.5°Cat760mmHg.
Formula Weight 280.07
Percent Purity ≥98%
Chemical Name or Material Pentafluorophenyl trifluoroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'