Läs mer
Pentamidine Diisetionate, LoGiCal
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pentamidine Diisetionate |
| CAS | 140-64-7 |
| Analyt- eller komponentnamn | Pentamidine Diisetionate |
| Frakt skick | Room Temperature |
| Molekylformel | C19 H24 N4 O2 . 2 C2 H6 O4 S |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Pentamidine diisetionate, 4,4'-[Pentane-1,5-diylbis(oxy)]dibenzamidine di(2-hydroxyethanesulfonate), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-[1,5-pentanediylbis(oxy)]bis[benzenecarboximidamide] (2:1), Benzamidine, 4,4'-(pentamethylenedioxy)di-, diisethionate (6CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 4,4'-(pentamethylenedioxy)dibenzamidine (2:1) (8CI), Benzenecarboximidamide, 4,4'-[1,5-pentanediylbis(oxy)]bis-, bis(2-hydroxyethanesulfonate) (9CI), 1,5-Bis(4-amidinophenoxy)pentane diisethionate, 2512RP, 4,4'-Diamidino-α-diphenoxypentane diisethionate, 4,4'-Diamidinodiphenoxypentane di(β-hydroxyethanesulfonate), Aeropent, Banambax, Diamidine, Lomidin, Lomidine, M & B 800, NebuPent, Pentacarinat, Pentam, Pentam 300, Pentamidine diisethionate, Pentamidine isethionate, Pentamidine isetionate, Pneumopent, RP 2512, WR 4931, p,p'-(Pentamethylenedioxy)dibenzamidine bis(β-hydroxyethanesulfonate) |
| InChI formel | InChI=1S/C19H24N4O2.2C2H6O4S/c20-18(21)14-4-8-16(9-5-14)24-12-2-1-3-13-25-17-10-6-15(7-11-17)19(22)23;2*3-1-2-7(4,5)6/h4-11H,1-3,12-13H2,(H3,20,21)(H3,22,23);2*3H,1-2H2,(H,4,5,6) |
| Lösningstyp | Neat |
| LEDER | NC(=N)c1ccc(OCCCCCOc2ccc(cc2)C(=N)N)cc1.OCCS(=O)(=O)O.OCCS(=O)(=O)O |
| Visa mer |