Läs mer
Pentamidine isethionate, MedChemExpress
Produktkod. 16484387
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16484387 50 mg
16478356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16484387 Leverantör Medchem Express Leverantörsnummer HYB0537B
MedChemExpress Pentamidine isethionate (MP-601205 isethionate) is an antimicrobial agent and interferes with DNA biosynthetics.
Pentamidine isethionate inhibits parasite Leishmania infantum with an IC50 of 2.5 μM. Pentamidine isethionate is a potent and selective protein tyrosine phosphatases (PTPases) and phosphatase of regenerating liver (PRL) inhibitor. Pentamidine isethionate has the potential for Gambian trypanosomiasis, antimony-resistant leishmaniasis, and Pneumocystis carinii pneumonia treatment. Antitumor and antibacterial activities.
Specifikationer
Kemiskt namn eller material Pentamidine isethionate
Fysisk form Solid
CAS 140-64-7
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C23H36N4O10S2
För användning med (applikation) COVID-19-immunoregulation
Synonym MP-601205 isethionate
Löslighetsinformation H2O : 100 mg/mL (168.73 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (168.73 mM; Need ultrasonic)
LEDER N=C(C1=CC=C(OCCCCCOC2=CC=C(C(N)=N)C=C2)C=C1)N.OCCS(=O)(O)=O.OCCS(=O)(O)=O
Molekylvikt (g/mol) 592.68
Formel vikt 592.68
Procent renhet 99.78%