Läs mer
pentan-2-amine hydrochloride, TRC
Produktkod. 30343719
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30343719 1 g
1 options
Produktkod. 30343719 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5106551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material pentan-2-amine hydrochloride
CAS 76716-21-7
Molekylformel C5H13N.HCl
Kvantitet 1 g
Synonym 2-Pentanamine, hydrochloride (1:1) (ACI), 2-Pentanamine, hydrochloride (9CI), Butylamine, 1-methyl-, hydrochloride (6CI), 2-Pentylamine hydrochloride, 2-Pentanamine hydrochloride, (±)-1-Methylbutylamine hydrochloride, 2-Aminopentane hydrochloride
InChI formel InChI=1S/C5H13N.ClH/c1-3-4-5(2)6;/h5H,3-4,6H2,1-2H3;1H
LEDER Cl.CCCC(C)N
IUPAC-namn pentan-2-amine;hydrochloride
Molekylvikt (g/mol) 123.62
Formel vikt 123.0815