Läs mer
Pentisomide, TRC
Produktkod. 30421939
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30421939 2.5 mg 2.5mg
30409570 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30421939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2756002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78833-03-1
Molekylformel C19 H33 N3 O
Molekylvikt (g/mol) 319.48
IUPAC-namn 2-[2-[di(propan-2-yl)amino]ethyl]-4-methyl-2-pyridin-2-ylpentanamide
LEDER CC(C)CC(CCN(C(C)C)C(C)C)(C(=O)N)c1ccccn1
Specifikationer
Kemiskt namn eller material Pentisomide
Kvantitet 2.5 mg
InChI formel InChI=1S/C19H33N3O/c1-14(2)13-19(18(20)23,17-9-7-8-11-21-17)10-12-22(15(3)4)16(5)6/h7-9,11,14-16H,10,12-13H2,1-6H3,(H2,20,23)
Formel vikt 319.2624