Läs mer
Pentyl Hexanoate, TRC
Produktkod. 30427932
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30427932 10 mg
1 options
Produktkod. 30427932 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP57088010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pentyl Hexanoate
CAS 540-07-8
Molekylformel C11H22O2
Kvantitet 10 mg
Synonym Amyl Caproate, Amyl Capronate, Amyl Hexanoate, n-Amyl Caproate, n-Pentyl Hexanoate, NSC 46119, NSC 53794, Pentyl Caproate
InChI formel InChI=1S/C11H22O2/c1-3-5-7-9-11(12)13-10-8-6-4-2/h3-10H2,1-2H3
LEDER O=C(OCCCCC)CCCCC
Molekylvikt (g/mol) 186.29