Läs mer
Pentylboronic Acid, TRC
Produktkod. 30418364
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30418364 5 g
1 options
Produktkod. 30418364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2797805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pentylboronic Acid
CAS 4737-50-2
Molekylformel C5H13BO2
Kvantitet 5 g
Synonym 1-Pentaneboronic Acid, Pentyl-Boronic Acid, NSC 524968, Pentylboronic Acid, B-Pentylboronic Acid
InChI formel InChI=1S/C5H13BO2/c1-2-3-4-5-6(7)8/h7-8H,2-5H2,1H3
LEDER CCCCCB(O)O
IUPAC-namn pentylboronic acid
Molekylvikt (g/mol) 115.97
Formel vikt 116.1009