Learn More
Thermo Scientific Chemicals Pepsin from Porcine Stomach
Derived from Porcine stomach
7860.00 SEK - 9699.00 SEK
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15875178
SDS View Documents Promotion Details
Thermo Scientific™
J20010.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15845328
SDS View Documents Promotion Details
Thermo Scientific Chemicals
J20015.A1
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.