Learn More
Thermo Scientific Chemicals Pepsin from Porcine Stomach
Derived from Porcine stomach
7860.00 SEK - 9699.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15875178
|
Thermo Scientific™
J20010.14 |
25 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15845328
|
Thermo Scientific Chemicals
J20015.A1 |
1 kg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.
RUO – Research Use Only