Läs mer
Invitrogen™ Peptidoglycan Monoclonal Antibody (3F6B3 (10H6))
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516509
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Treatment with strong acid (Gram positive bacteria) or with a detergent such as SDS (Gram-negative bacteria) may be necessary to expose the epitope.
Specifikationer
| Peptidoglycan | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with no preservative | |
| Mouse | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 3F6B3 (10H6) | |
| Unconjugated | |
| PG | |
| This antibody was raised against insoluble peptidoglycan obtained by TCA-heat and ethanol extraction of Streptococcus mutans BHT cells. | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |