Learn More
PER2 Mouse anti-Human, Unconjugated, Clone: 2E6C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67513-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16830186
  • 4721.67 SEK / 150µL
Estimated Shipment: 26-04-2024
to see stock.
Description
Description
The PER2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PER2. This antibody recognizes human, rat antigen. The PER2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PER2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot
2E6C3
Unconjugated
150 μL
8864
IgG1