Learn More
PER2 Mouse anti-Human, Unconjugated, Clone: 2E6C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67513-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The PER2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PER2. This antibody recognizes human, rat antigen. The PER2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PER2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Western Blot | |
2E6C3 | |
Unconjugated | |
150 μL | |
8864 | |
IgG1 |